CAS RN 100587-95-9
CAS RN 100587-95-9
IUPAC Name:
europium(3+) hydrate trinitrate
Synonyms:
europium(3+) hydrate trinitrate
Molecular Weight (g/mol):
355.99
Molecular Formula:
EuH2N3O10
InChi Key:
XLGOHVJDOSTECY-UHFFFAOYSA-N
SMILES:
O.[Eu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
1
–
1
of
1
results
1
Europium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
CAS: 100587-95-9 Molecular Formula: EuN3O9 Molecular Weight (g/mol): 337.98 MDL Number: MFCD00149700 InChI Key: GAGGCOKRLXYWIV-UHFFFAOYSA-N Synonym: europium iii nitrate hydrate,acmc-1c6y6,eu.3no3.h2o,europium trinitrate monohydrate,europium 3+ hydrate trinitrate,europium 3+ ion hydrate trinitrate,nitric acid,europium 3+ salt, hydrate 9ci PubChem CID: 57346089 SMILES: [Eu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O