accessibility menu, dialog, popup

Test

CAS RN 1005-38-5

CAS RN 1005-38-5

IUPAC Name: 6-chloro-2-(methylsulfanyl)pyrimidin-4-amine
Synonyms: 6-chloro-2-(methylsulfanyl)pyrimidin-4-amine
Molecular Weight (g/mol): 175.63
Molecular Formula: C5H6ClN3S
InChi Key: ISUXMAHVLFRZQU-UHFFFAOYSA-N
SMILES: CSC1=NC(N)=CC(Cl)=N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)