accessibility menu, dialog, popup

Test

CAS RN 10045-52-0

CH3OSN

CAS RN 10045-52-0

IUPAC Name: 1-(2-phenyl-1,3-thiazol-4-yl)ethan-1-one
Synonyms: 1-(2-phenyl-1,3-thiazol-4-yl)ethanone
Molecular Weight (g/mol): 203.26
Molecular Formula: C11H9NOS
InChi Key: ZOOGZFPRAKXWKI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CSC(=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results