CAS RN 10043-52-4
CAS RN 10043-52-4
Calcium chloride, 93%, extra pure, powder, anhydrous, Thermo Scientific Chemicals
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 SMILES: [Cl-].[Cl-].[Ca++]
Calcium chloride, dried, powder, 97%, Thermo Scientific Chemicals
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 IUPAC Name: calcium;dichloride SMILES: [Cl-].[Cl-].[Ca++]
Calcium Chloride, Extra Pure, SLR, Fused, Granular, Fisher Chemical™
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: 10903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L IUPAC Name: calcium dichloride SMILES: [Cl-].[Cl-].[Ca++]
Calcium chloride, Anhydrous, powder, ≥97%, Honeywell Fluka™
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 SMILES: [Cl-].[Cl-].[Ca++]
Calcium chloride, anhydrous, 93%, Thermo Scientific Chemicals
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 SMILES: [Cl-].[Cl-].[Ca++]
Calcium chloride, 1M aq. soln., Thermo Scientific Chemicals
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 IUPAC Name: calcium dichloride SMILES: [Cl-].[Cl-].[Ca++]
Calcium chloride, 96%, for biochemistry, anhydrous, Thermo Scientific Chemicals
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 IUPAC Name: calcium dichloride SMILES: [Cl-].[Cl-].[Ca++]
Calcium chloride, anhydrous, porous, 93%, Thermo Scientific Chemicals
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 SMILES: [Cl-].[Cl-].[Ca++]
Calcium chloride, 96%, for analysis, granules, Thermo Scientific Chemicals
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 IUPAC Name: calcium dichloride SMILES: [Cl-].[Cl-].[Ca++]
Calcium chloride, anhydrous, ACS, 96.0% min, Thermo Scientific Chemicals
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 IUPAC Name: calcium dichloride SMILES: [Cl-].[Cl-].[Ca++]
Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 SMILES: [Cl-].[Cl-].[Ca++]
Calcium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.98 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L Synonym: calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal PubChem CID: 5284359 ChEBI: CHEBI:3312 IUPAC Name: calcium dichloride SMILES: [Cl-].[Cl-].[Ca++]
- 1
- 2