CAS RN 10041-02-8
CAS RN 10041-02-8
IUPAC Name:
4-(1H-imidazol-1-yl)phenol
Synonyms:
4-(1H-imidazol-1-yl)phenol
Molecular Weight (g/mol):
160.18
Molecular Formula:
C9H8N2O
InChi Key:
CYKCUAPYWQDIKR-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1)N1C=CN=C1
1
–
3
of
3
results
1
4-(1-Imidazolyl)phenol, 97%, Thermo Scientific Chemicals
CAS: 10041-02-8 Molecular Formula: C9H8N2O Molecular Weight (g/mol): 160.18 MDL Number: MFCD00005281 InChI Key: CYKCUAPYWQDIKR-UHFFFAOYSA-N Synonym: 4-imidazol-1-yl phenol,4-1h-imidazol-1-yl phenol,4-imidazol-1-yl-phenol,4-1-imidazolyl phenol,1-p-hydroxyphenyl imidazole,phenol, 4-1h-imidazol-1-yl,1-4-hydroxyphenyl imidazole,4-imidazolylphenol,4‐ 1h‐imidazol‐1‐yl phenol compound 1,p-1-imidazolyl phenol PubChem CID: 82315 IUPAC Name: 4-imidazol-1-ylphenol SMILES: OC1=CC=C(C=C1)N1C=CN=C1