accessibility menu, dialog, popup

Test

CAS RN 1004-36-0

CH3OH3CO

CAS RN 1004-36-0

IUPAC Name: 2,6-dimethyl-4H-pyran-4-one
Synonyms: gamma-2,6-dimethylpyrone
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: VSYFZULSKMFUJJ-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C=C(C)O1
Boiling Point 
  • (6)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)