accessibility menu, dialog, popup

Test

CAS RN 10031-43-3

CuONOOONOO

CAS RN 10031-43-3

IUPAC Name: copper(2+) dinitrate
Synonyms: copper(II) nitrate
Molecular Weight (g/mol): 187.55
Molecular Formula: CuN2O6
InChi Key: XTVVROIMIGLXTD-UHFFFAOYSA-N
SMILES: [Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (5)
Physical Form 
  • (4)
  • (4)
Grade 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (5)
  • (4)
  • (4)
  • (4)
  • (2)