accessibility menu, dialog, popup

Test

CAS RN 10030-85-0

CH3OOHOHHOHO

CAS RN 10030-85-0

IUPAC Name: (2R,3R,4R,5R,6S)-6-methyloxane-2,3,4,5-tetrol
Synonyms: α-L-rhamnopyranose α-L-mannomethylose α-6-deoxy-L-mannose α-L-rhamnose α-L-rha
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: SHZGCJCMOBCMKK-HGVZOGFYSA-N
SMILES: C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
Quantity 
  • (4)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (6)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (6)
  • (3)
  • (1)