CAS RN 1003-04-9
CAS RN 1003-04-9
IUPAC Name:
thiolan-3-one
Synonyms:
thiolan-3-one
Molecular Weight (g/mol):
102.15
Molecular Formula:
C4H6OS
InChi Key:
DSXFPRKPFJRPIB-UHFFFAOYSA-N
SMILES:
O=C1CCSC1
1
–
2
of
2
results
2
Tetrahydrothiophen-3-one, 97%, Thermo Scientific Chemicals
CAS: 1003-04-9 Molecular Formula: C4H6OS Molecular Weight (g/mol): 102.15 MDL Number: MFCD00005412 InChI Key: DSXFPRKPFJRPIB-UHFFFAOYSA-N Synonym: tetrahydrothiophen-3-one,3-thiophanone,dihydrothiophen-3 2h-one,dihydro-3 2h-thiophenone,3-thiacyclopentanone,3-oxotetrahydrothiophene,3 2h-thiophenone, dihydro,3-tetrahydrothiophenone,tetrahydrothiophene-3-one,3-oxo-2,3,4,5-tetrahydrothiophene PubChem CID: 61252 IUPAC Name: thiolan-3-one SMILES: O=C1CCSC1