accessibility menu, dialog, popup

Test

CAS RN 10026-06-9

H2OH2OH2OClClClClH2OH2OSn

CAS RN 10026-06-9

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 354.62
Molecular Formula: Cl4H14O5Sn
InChi Key: XOWGNBAIXFJDCN-UHFFFAOYSA-N
SMILES: O.O.O.O.O.Cl.Cl.Cl.Cl.[Sn]
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)
  • (2)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)