accessibility menu, dialog, popup

Test

CAS RN 10026-03-6

ClSeClClCl

CAS RN 10026-03-6

IUPAC Name: tetrachloro-λ⁴-selane
Synonyms: selenium tetrachloride
Molecular Weight (g/mol): 220.77
Molecular Formula: Cl4Se
InChi Key: LNBXMNQCXXEHFT-UHFFFAOYSA-N
SMILES: Cl[Se](Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results