accessibility menu, dialog, popup

Test

CAS RN 10025-76-0

ClClClEu

CAS RN 10025-76-0

IUPAC Name: europium(3+) trichloride
Synonyms: europium(III) chloride
Molecular Weight (g/mol): 258.31
Molecular Formula: Cl3Eu
InChi Key: NNMXSTWQJRPBJZ-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Eu+3]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results