CAS RN 10025-74-8
CAS RN 10025-74-8
IUPAC Name:
dysprosium(3+) trichloride
Synonyms:
dysprosium(III) chloride
Molecular Weight (g/mol):
268.85
Molecular Formula:
Cl3Dy
InChi Key:
BOXVSFHSLKQLNZ-UHFFFAOYSA-K
SMILES:
[Cl-].[Cl-].[Cl-].[Dy+3]
1
–
1
of
1
results
1
Dysprosium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals
CAS: 10025-74-8 Molecular Formula: Cl3Dy Molecular Weight (g/mol): 268.85 MDL Number: MFCD00010984 InChI Key: BOXVSFHSLKQLNZ-UHFFFAOYSA-K IUPAC Name: dysprosium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Dy+3]