accessibility menu, dialog, popup

Test

CAS RN 100036-64-4

FFFO

CAS RN 100036-64-4

IUPAC Name: 3'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde
Synonyms: 3'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde
Molecular Weight (g/mol): 250.22
Molecular Formula: C14H9F3O
InChi Key: WRWNNARTYPYHEC-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)C1=CC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results