accessibility menu, dialog, popup

Test

CAS RN 100020-57-3

CH3SNNNH2NNOOPOOHOPOHOOPOHOOHOHOHNaNaNaNa

CAS RN 100020-57-3

IUPAC Name: [({[({5-[6-amino-2-(methylsulfanyl)-9H-purin-9-yl]-3,4-dihydroxyoxolan-2-yl}methoxy)(hydroxy)phosphoryl]oxy}(hydroxy)phosphoryl)oxy]phosphonic acid tetrasodium
Synonyms: [({5-[6-amino-2-(methylsulfanyl)purin-9-yl]-3,4-dihydroxyoxolan-2-yl}methoxy(hydroxy)phosphoryl)oxy(hydroxy)phosphoryl]oxyphosphonic acid tetrasodium
Molecular Weight (g/mol): 645.23
Molecular Formula: C11H18N5Na4O13P3S
InChi Key: ODWHHSGFUKWMOE-UHFFFAOYNA-N
SMILES: [Na].[Na].[Na].[Na].CSC1=NC(N)=C2N=CN(C3OC(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)C(O)C3O)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results