accessibility menu, dialog, popup

Test

CAS RN 100-97-0

NNNN

CAS RN 100-97-0

IUPAC Name: 1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
Synonyms: formin caswell no. 482 S 4 (heterocycle) esametilentetramina hexamethylenamine hexamethylenetetraamine cystamin urotropine hexaform ammoform
Molecular Weight (g/mol): 140.19
Molecular Formula: C6H12N4
InChi Key: VKYKSIONXSXAKP-UHFFFAOYSA-N
SMILES: C1N2CN3CN1CN(C2)C3
Quantity 
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
Physical Form 
  • (3)
  • (3)
  • (9)
Percent Purity 
  • (7)
  • (2)
  • (3)
Grade 
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (1)
  • (5)
  • (6)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (3)
  • (9)
  • (7)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)