man

Test

CAS RN 100-86-7

H3CH3COH

CAS RN 100-86-7

IUPAC Name: 2-methyl-1-phenylpropan-2-ol
Synonyms: dimethylbenzyl carbinol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: RIWRBSMFKVOJMN-UHFFFAOYSA-N
SMILES: CC(C)(O)CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results