man

Test

CAS RN 100-83-4

OHO

CAS RN 100-83-4

IUPAC Name: 3-hydroxybenzaldehyde
Synonyms: 3-hydroxybenzaldehyde
Molecular Weight (g/mol): 122.12
Molecular Formula: C7H6O2
InChi Key: IAVREABSGIHHMO-UHFFFAOYSA-N
SMILES: OC1=CC=CC(C=O)=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)