accessibility menu, dialog, popup

Test

CAS RN 100-82-3

H2NF

CAS RN 100-82-3

IUPAC Name: 1-(3-fluorophenyl)methanamine
Synonyms: 1-(3-fluorophenyl)methanamine
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: QVSVMNXRLWSNGS-UHFFFAOYSA-N
SMILES: NCC1=CC=CC(F)=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)