accessibility menu, dialog, popup

Test

CAS RN 100-72-1

CAS RN 100-72-1

IUPAC Name: (oxan-2-yl)methanol
Synonyms: tetrahydropyran-2-methanol
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: ROTONRWJLXYJBD-UHFFFAOYNA-N
SMILES: OCC1CCCCO1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)