man

Test

CAS RN 100-69-6

H2CN

CAS RN 100-69-6

IUPAC Name: 2-ethenylpyridine
Synonyms: 2-vinylpyridine
Molecular Weight (g/mol): 105.14
Molecular Formula: C7H7N
InChi Key: KGIGUEBEKRSTEW-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)