accessibility menu, dialog, popup

Test

CAS RN 100-58-3

CAS RN 100-58-3

IUPAC Name: phenylmagnesium bromide
Synonyms: phenylmagnesium bromide
Molecular Weight (g/mol): 181.32
Molecular Formula: C6H5BrMg
InChi Key:
SMILES: [Mg++].[Br-].c1cc[cH-]cc1
Quantity 
  • (4)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (4)
  • (1)
  • (1)
  • (3)