man

Test

CAS RN 100-52-7

O

CAS RN 100-52-7

IUPAC Name: benzaldehyde
Synonyms: benzaldehyde caswell no. 076 artificial essential oil of almond synthetic bitter almond oil benzaldehyde ffc BEZ bitter almond oil, synthetic benzaldehyde (product) benzoic acid aldehyde phenylmethanal benzenecarboxaldehyde
Molecular Weight (g/mol): 106.12
Molecular Formula: C7H6O
InChi Key: HUMNYLRZRPPJDN-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (6)
Molecular Weight (g/mol) 
  • (6)
  • (1)
Grade 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results