accessibility menu, dialog, popup

Test

CAS RN 100-48-1

NN

CAS RN 100-48-1

IUPAC Name: pyridine-4-carbonitrile
Synonyms: 4-cyanopyridine isonicotinic acid nitrile
Molecular Weight (g/mol): 104.11
Molecular Formula: C6H4N2
InChi Key: GPHQHTOMRSGBNZ-UHFFFAOYSA-N
SMILES: N#CC1=CC=NC=C1
Boiling Point 
  • (4)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (1)
  • (1)
  • (4)
  • (5)
  • (5)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)