man

Test

CAS RN 100-21-0

HOOOHO

CAS RN 100-21-0

IUPAC Name: benzene-1,4-dicarboxylic acid
Synonyms: terephthalic acid P-phthelate TA-33MP tephthol kyselina terftalova TPA para-phthalic acid p-phthalate p-phthalic acid P-phthelic acid
Molecular Weight (g/mol): 166.13
Molecular Formula: C8H6O4
InChi Key: KKEYFWRCBNTPAC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (6)
  • (5)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)