man

Test

CAS RN 100-07-2

H3COClO

CAS RN 100-07-2

IUPAC Name: 4-methoxybenzoyl chloride
Synonyms: benzoyl chloride, 4-methoxy-
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: MXMOTZIXVICDSD-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(Cl)=O
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)